Få mere at vide
4-Fluorobenzyl-alpha,alpha-d2 Chloride, CDN
Artikelnummer. 30320933
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320933 0.25 g
30310589 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320933 Leverandør CDN Leverandørnr. CDND60770.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Fluorobenzyl Chloride-alpha,alpha-D2
CAS 1219804-18-8
Molekylær formel C7 D2 H4 Cl F
Mængde 0.25 g
Synonym 4-Fluorobenzyl-α,α-d2 Chloride, 4-Fluorobenzyl Chloride-d2, 4-Fluorobenzyl Chloride-α,α-D2, 1-(Chloromethyl-d2)-4-fluorobenzene, α-Chloro-4-fluorotoluene-α,α-d2
InChI formel InChI=1 S/C7H6ClF/c8-5-6-1-3-7(9)4-2-6/h1-4 H,5H2/i5D2
SMIL [2 H]C([2 H])(Cl)c1ccc(F)cc1
IUPAC navn 1-[chloro(dideuterio)methyl]-4-fluorobenzene
Molekylvægt (g/mol) 146.5863
Formel vægt 146.0268 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity
Vis mere Vis mindre
Produkttitel
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.