Få mere at vide
4-Fluorobenzyl-alpha,alpha-d2 Chloride, CDN
Artikelnummer. 30310589
Skift visning
Mængde:
0.25 g
0.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310589 0.5 g
30320933 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310589 Leverandør CDN Leverandørnr. CDND60770.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Fluorobenzyl Chloride-alpha,alpha-D2
CAS 1219804-18-8
Molekylær formel C7 D2 H4 Cl F
Mængde 0.5 g
Synonym 4-Fluorobenzyl-α,α-d2 Chloride, 4-Fluorobenzyl Chloride-d2, 4-Fluorobenzyl Chloride-α,α-D2, 1-(Chloromethyl-d2)-4-fluorobenzene, α-Chloro-4-fluorotoluene-α,α-d2
InChI formel InChI=1 S/C7H6ClF/c8-5-6-1-3-7(9)4-2-6/h1-4 H,5H2/i5D2
SMIL [2 H]C([2 H])(Cl)c1ccc(F)cc1
IUPAC navn 1-[chloro(dideuterio)methyl]-4-fluorobenzene
Molekylvægt (g/mol) 146.5863
Formel vægt 146.0268 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity