Få mere at vide
4-Fluorobenzamide-D4, TRC
Artikelnummer. 30406531
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30406531 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406531 Leverandør Toronto Research Chemicals Leverandørnr. TRCF58827210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Fluorobenzamide-D4
CAS 1100750-65-9
Molekylær formel C7H2D4FNO
Mængde 10 mg
InChI formel InChI=1S/C7H6FNO/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H2,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C(=O)N)c([2H])c([2H])c1F
IUPAC navn 2,3,5,6-tetradeuterio-4-fluorobenzamide
Molekylvægt (g/mol) 143.15
Formel vægt 143.0684