Få mere at vide
4-Fluorobenzaldehyde-2,3,5,6-d4, TRC
Artikelnummer. 30406740
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30406740 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406740 Leverandør Toronto Research Chemicals Leverandørnr. TRCF588517500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Fluorobenzaldehyde-2,3,5,6-d4
CAS 93111-27-4
Molekylær formel C7 D4 H F O
Mængde 500 mg
Synonym p-Fluorobenzaldehyde-d4, p-Fluorobenzenecarboxaldehyde-d4
InChI formel InChI=1S/C7H5FO/c8-7-3-1-6(5-9)2-4-7/h1-5H/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(C=O)c([2H])c([2H])c1F
IUPAC navn 2,3,5,6-tetradeuterio-4-fluorobenzaldehyde
Molekylvægt (g/mol) 128.14
Formel vægt 128.0575