missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)-2-(trifluoromethyl)benzamide, 95%

Brand:  Alfa Aesar™ H65723.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469856

  • 1908.81 DKK / 1g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27975810

Specifications

4-Fluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)-2-(trifluoromethyl)benzamide
MFCD27975810
Solid
1g
337.32
95%
Videos
SDS