missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-DL-α-phenylglycine, 98+%, Thermo Scientific™

Catalog Number p-3212069
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7292-73-1
C8H8FNO2
169.16
MFCD00066444,MFCD00042727
JKFYKCYQEWQPTM-UHFFFAOYNA-N
2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-dl-phenylglycine, 4-fluoro-dl-alpha-phenylglycine, dl-4-fluorophenylglycine, amino 4-fluorophenyl acetic acid, 4-fluorophenylglycine, dl-4-fluorophenyl glycine, amino-4-fluoro-phenyl-acetic acid, dl-alpha-4-fluorophenyl-glycine, benzeneacetic acid, alpha-amino-4-fluoro
98313
2-amino-2-(4-fluorophenyl)acetic acid
NC(C(O)=O)C1=CC=C(F)C=C1
This item is not returnable. View return policy
7292-73-1
C8H8FNO2
169.16
MFCD00066444,MFCD00042727
JKFYKCYQEWQPTM-UHFFFAOYNA-N
2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-dl-phenylglycine, 4-fluoro-dl-alpha-phenylglycine, dl-4-fluorophenylglycine, amino 4-fluorophenyl acetic acid, 4-fluorophenylglycine, dl-4-fluorophenyl glycine, amino-4-fluoro-phenyl-acetic acid, dl-alpha-4-fluorophenyl-glycine, benzeneacetic acid, alpha-amino-4-fluoro
98313
2-amino-2-(4-fluorophenyl)acetic acid
NC(C(O)=O)C1=CC=C(F)C=C1
CAS Min % 98.0
CAS Max % 100.0
Melting Point 300°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H4CH(NH2)COOH
Quantity 1 g
Formula Weight 169.15
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 4-Fluoro-DL-α-phenylglycine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'