missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog Number 11485786
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
67515-60-0
C8H4F4O
192.113
MFCD00061314
BIUDHHGROGJSHN-UHFFFAOYSA-N
4-fluoro-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-m-tolualdehyde, pubchem1462, acmc-209nzm, ksc359a9l, 4-fluoro-3-trifluoromethyl
522269
4-fluoro-3-(trifluoromethyl)benzaldehyde
C1=CC(=C(C=C1C=O)C(F)(F)F)F
This item is not returnable. View return policy
67515-60-0
C8H4F4O
192.113
MFCD00061314
BIUDHHGROGJSHN-UHFFFAOYSA-N
4-fluoro-3-trifluoromethyl benzaldehyde, 3-trifluoromethyl-4-fluorobenzaldehyde, benzaldehyde, 4-fluoro-3-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-m-tolualdehyde, pubchem1462, acmc-209nzm, ksc359a9l, 4-fluoro-3-trifluoromethyl
522269
4-fluoro-3-(trifluoromethyl)benzaldehyde
C1=CC(=C(C=C1C=O)C(F)(F)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.408
Boiling Point 178°C
Flash Point 76°C (168°F)
Refractive Index 1.457
Quantity 1 g
Beilstein 1958528
Sensitivity Air Sensitive
Formula Weight 192.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'