missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-3-methylbenzenesulfonyl chloride, 97%

Brand:  Alfa Aesar™ H64110.03

Additional Details : CAS Number : 629672-19-1 Weight : 0.00100kg Transport : UN number : 3261 Chem class : 8 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 15471159

  • 416.15 DKK / 1g


Estimated Shipment date
from Supplier 03-07-2020
Add to basket

Specifications

Chemical Identifiers

629672-19-1
208.631
ALEQALYEZFNWOP-UHFFFAOYSA-N
2782758
CC1=C(C=CC(=C1)S(=O)(=O)Cl)F
C7H6ClFO2S
MFCD01365818
4-fluoro-3-methyl-benzenesulfonyl chloride, 4-fluoro-3-methylbenzenesulfonylchloride, 4-fluoro-3-methylbenzene-1-sulfonyl chloride, 4-fluoro-3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro-3-methyl, chloro 4-fluoro-3-methylphenyl sulfone, ablock ab-12-8863, 5-chlorosulphonyl-2-fluorotoluene, 4-fluro-3-methylphenyl-sulfonylchloride, 4-fluoro-3-methylphenyl-sulfonylchloride
4-fluoro-3-methylbenzenesulfonyl chloride

Specifications

4-Fluoro-3-methylbenzenesulfonyl chloride
Moisture sensitive
1g
C7H6ClFO2S
UN3261
4-fluoro-3-methyl-benzenesulfonyl chloride, 4-fluoro-3-methylbenzenesulfonylchloride, 4-fluoro-3-methylbenzene-1-sulfonyl chloride, 4-fluoro-3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro-3-methyl, chloro 4-fluoro-3-methylphenyl sulfone, ablock ab-12-8863, 5-chlorosulphonyl-2-fluorotoluene, 4-fluro-3-methylphenyl-sulfonylchloride, 4-fluoro-3-methylphenyl-sulfonylchloride
CC1=C(C=CC(=C1)S(=O)(=O)Cl)F
208.631
208.63
97%
>110°C (230°F)
35°C to 40°C
629672-19-1
MFCD01365818
3254603
ALEQALYEZFNWOP-UHFFFAOYSA-N
4-fluoro-3-methylbenzenesulfonyl chloride
2782758
Solid
Videos
SDS Documents