missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-7055509
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
629672-19-1
C7H6ClFO2S
208.63
MFCD01365818
ALEQALYEZFNWOP-UHFFFAOYSA-N
4-fluoro-3-methyl-benzenesulfonyl chloride, 4-fluoro-3-methylbenzenesulfonylchloride, 4-fluoro-3-methylbenzene-1-sulfonyl chloride, 4-fluoro-3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro-3-methyl, chloro 4-fluoro-3-methylphenyl sulfone, ablock ab-12-8863, 5-chlorosulphonyl-2-fluorotoluene, 4-fluro-3-methylphenyl-sulfonylchloride, 4-fluoro-3-methylphenyl-sulfonylchloride
2782758
CC1=C(F)C=CC(=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
629672-19-1
C7H6ClFO2S
208.63
MFCD01365818
ALEQALYEZFNWOP-UHFFFAOYSA-N
4-fluoro-3-methyl-benzenesulfonyl chloride, 4-fluoro-3-methylbenzenesulfonylchloride, 4-fluoro-3-methylbenzene-1-sulfonyl chloride, 4-fluoro-3-methylbenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro-3-methyl, chloro 4-fluoro-3-methylphenyl sulfone, ablock ab-12-8863, 5-chlorosulphonyl-2-fluorotoluene, 4-fluro-3-methylphenyl-sulfonylchloride, 4-fluoro-3-methylphenyl-sulfonylchloride
2782758
CC1=C(F)C=CC(=C1)S(Cl)(=O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 35°C to 40°C
Flash Point >110°C (230°F)
Quantity 250 mg
UN Number UN3261
Beilstein 3254603
Sensitivity Moisture sensitive
IUPAC Name 4-fluoro-3-methylbenzenesulfonyl chloride
Formula Weight 208.63
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Fluoro-3-methylbenzenesulfonyl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'