missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylaniline, 97%, Thermo Scientific Chemicals

Catalog Number p-7022618
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
452-69-7
C7H8FN
125.15
MFCD00025294
NYMDPDNETOLVBS-UHFFFAOYSA-N
2-fluoro-5-aminotoluene, 4-fluoro-m-toluidine, 5-amino-2-fluorotoluene, 4-fluoro-3-methylbenzenamine, 4-fluoro-3-methylphenylamine, benzenamine, 4-fluoro-3-methyl, 4-fluoro-3-methyl aniline, 4-fluoro-3-methyl-aniline, 4-fluor-3-methylanilin, pubchem1513
67981
4-fluoro-3-methylaniline
CC1=CC(N)=CC=C1F
This item is not returnable. View return policy
452-69-7
C7H8FN
125.15
MFCD00025294
NYMDPDNETOLVBS-UHFFFAOYSA-N
2-fluoro-5-aminotoluene, 4-fluoro-m-toluidine, 5-amino-2-fluorotoluene, 4-fluoro-3-methylbenzenamine, 4-fluoro-3-methylphenylamine, benzenamine, 4-fluoro-3-methyl, 4-fluoro-3-methyl aniline, 4-fluoro-3-methyl-aniline, 4-fluor-3-methylanilin, pubchem1513
67981
4-fluoro-3-methylaniline
CC1=CC(N)=CC=C1F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 32°C to 36°C
Density 1.12
Boiling Point 85°C to 86°C (9 mmHg)
Flash Point 95°C (203°F)
Odor Characteristic
Refractive Index 1.537
Quantity 5 g
UN Number UN2811
Beilstein 2935561
Formula Weight 125.15
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'