missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62385.MD

Additional Details : Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15437366

  • 1261.00 DKK / 250mg

Estimated Shipment
08-04-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

872459-87-5
280.102
methyl 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-fluoro-3-methoxycarbonylphenylboronic acid pinacol ester, benzoic acid, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester, 4-fluoro-3-methoxycorbonyl benzeneboronic acid pinacol ester, 4-fluoro-3-methoxycorbonyl phenylboronic acid pinacol ester, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid, methyl ester
methyl 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
C14H18BFO4
PILMRJXFJLIVFU-UHFFFAOYSA-N
57880558
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C(=O)OC

Specifications

4-Fluoro-3-(methoxycarbonyl)benzeneboronic acid pinacol ester
872459-87-5
methyl 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 4-fluoro-3-methoxycarbonylphenylboronic acid pinacol ester, benzoic acid, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester, 4-fluoro-3-methoxycorbonyl benzeneboronic acid pinacol ester, 4-fluoro-3-methoxycorbonyl phenylboronic acid pinacol ester, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid, methyl ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C(=O)OC
280.102
236.09
96%
250mg
C14H18BFO4
PILMRJXFJLIVFU-UHFFFAOYSA-N
methyl 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
57880558
Solid
Videos
SDS Documents