missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)cinnamic acid, 97+%, Thermo Scientific™

Catalog Number 11489837
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
243977-21-1
C10H6F4O2
234.15
MFCD00236297
OREHSCSLACWBTF-DUXPYHPUSA-N
4-fluoro-2-trifluoromethyl cinnamic acid, 2e-3-4-fluoro-2-trifluoromethyl phenyl prop-2-enoic acid, 3-4-fluoro-2-trifluoromethyl phenyl acrylic acid, pubchem8430, 3-4-fluoro-2-trifluoromethyl-phenyl-acrylic acid, e-3-4-fluoro-2-trifluoromethyl phenyl acrylic acid, e-3-4-fluoro-2-trifluoromethyl phenyl-2-propenoic acid, e-3-4-fluoro-2-trifluoromethyl phenyl prop-2-enoic acid, e-3-4-fluoranyl-2-trifluoromethyl phenyl prop-2-enoic acid
5702896
(E)-3-[4-fluoro-2-(trifluoromethyl)phenyl]prop-2-enoic acid
C1=CC(=C(C=C1F)C(F)(F)F)C=CC(=O)O
This item is not returnable. View return policy
243977-21-1
C10H6F4O2
234.15
MFCD00236297
OREHSCSLACWBTF-DUXPYHPUSA-N
4-fluoro-2-trifluoromethyl cinnamic acid, 2e-3-4-fluoro-2-trifluoromethyl phenyl prop-2-enoic acid, 3-4-fluoro-2-trifluoromethyl phenyl acrylic acid, pubchem8430, 3-4-fluoro-2-trifluoromethyl-phenyl-acrylic acid, e-3-4-fluoro-2-trifluoromethyl phenyl acrylic acid, e-3-4-fluoro-2-trifluoromethyl phenyl-2-propenoic acid, e-3-4-fluoro-2-trifluoromethyl phenyl prop-2-enoic acid, e-3-4-fluoranyl-2-trifluoromethyl phenyl prop-2-enoic acid
5702896
(E)-3-[4-fluoro-2-(trifluoromethyl)phenyl]prop-2-enoic acid
C1=CC(=C(C=C1F)C(F)(F)F)C=CC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 181°C to 183°C
Quantity 1 g
Formula Weight 234.15
Percent Purity ≥97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)cinnamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'