missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrotoluene, 98%, Thermo Scientific™

Catalog Number p-7027704
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
446-10-6
C7H6FNO2
155.128
MFCD00007200
SKWTUNAAJNDEIK-UHFFFAOYSA-N
4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482
67965
4-fluoro-1-methyl-2-nitrobenzene
CC1=C(C=C(C=C1)F)[N+](=O)[O-]
This item is not returnable. View return policy
446-10-6
C7H6FNO2
155.128
MFCD00007200
SKWTUNAAJNDEIK-UHFFFAOYSA-N
4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482
67965
4-fluoro-1-methyl-2-nitrobenzene
CC1=C(C=C(C=C1)F)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 25°C to 27°C
Density 1.26
Boiling Point 137°C to 139°C (83 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.523
Quantity 5 g
UN Number UN2811
Beilstein 1946051
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 4-Fluoro-2-nitrotoluene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'