Learn More
4-Fluoro-2-methoxyaniline, 95%, Thermo Scientific Chemicals
Catalog Number 15458558
Click to view available options
Quantity:
250 mg
1 g
5 g
450-91-9
C7H8FNO
141.15
MFCD00077536
BNRRMRUVYDETQC-UHFFFAOYSA-N
2-methoxy-4-fluoroaniline, 2-amino-5-fluoroanisole, 4-fluoro-2-methoxybenzenamine, benzenamine, 4-fluoro-2-methoxy, 2-amino-5-fluoroanisole 4-fluoro-2-methoxyaniline, 4-fluoro-o-anisidine, acmc-1aegy, 4-fluoro-2-methoxy-aniline, ksc798g5t, 4-fluoro-2-methoxyphenylamine
13532228
4-fluoro-2-methoxyaniline
COC1=C(N)C=CC(F)=C1
450-91-9
C7H8FNO
141.15
MFCD00077536
BNRRMRUVYDETQC-UHFFFAOYSA-N
2-methoxy-4-fluoroaniline, 2-amino-5-fluoroanisole, 4-fluoro-2-methoxybenzenamine, benzenamine, 4-fluoro-2-methoxy, 2-amino-5-fluoroanisole 4-fluoro-2-methoxyaniline, 4-fluoro-o-anisidine, acmc-1aegy, 4-fluoro-2-methoxy-aniline, ksc798g5t, 4-fluoro-2-methoxyphenylamine
13532228
4-fluoro-2-methoxyaniline
COC1=C(N)C=CC(F)=C1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2811
Formula Weight 141.14
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Fluoro-2-methoxyaniline