Få mere at vide
4-Ethylaniline-d11, CDN
Artikelnummer. 30321224
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321224 0.5 g 0.5g
30310509 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321224 Leverandør CDN Leverandørnr. CDND60360.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Ethylaniline-D11
CAS 1219802-96-6
Molekylær formel C8 D11 N
Mængde 0.5 g
Synonym 4-(Ethyl-d5)benzen-1,2,5,6-d4-amine-d2, 4-Aminoethylbenzene-d11, 4-Ethylaniline-d11, 4-Ethylaniline-D11, p-Ethylaniline-d11
InChI formel InChI=1 S/C8H11N/c1-2-7-3-5-8(9)6-4-7/h3-6 H,2,9H2,1H3/i1D3,2D2,3 D,4 D,5 D,6 D/hD2
SMIL [2 H]N([2 H])c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn N,N,2,3,5,6-hexadeuterio-4-(1,1,2,2,2-pentadeuterioethyl)aniline
Molekylvægt (g/mol) 132.2474
Formel vægt 132.1582 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity