Få mere at vide
4-Ethyl-2-pyridinecarbonitrile-d5, TRC
Artikelnummer. 30370558
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30370558 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370558 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9255472.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Ethyl-2-pyridinecarbonitrile-d5
CAS 1330052-70-4
Molekylær formel C8H3D5N2
Mængde 2.5 mg
Synonym 2-Cyano-4-ethylpyridine-d5
InChI formel InChI=1S/C8H8N2/c1-2-7-3-4-10-8(5-7)6-9/h3-5H,2H2,1H3/i2D2,3D,4D,5D
SMIL [2H]c1nc(C#N)c([2H])c(c1[2H])C([2H])([2H])C
IUPAC navn 3,5,6-trideuterio-4-(1,1-dideuterioethyl)pyridine-2-carbonitrile
Molekylvægt (g/mol) 137.19
Formel vægt 137.1001