Få mere at vide
4-Ethoxy-3-nitropyridine hydrochloride, 97%, Thermo Scientific™
Artikelnummer. 11303676
Mængde:
5 g
25 g
Pakningsstørrelse:
25g
5g
Artikelnummer. 11303676
Brand: Thermo Scientific Alfa Aesar L14743.06
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Ethoxy-3-nitropyridine hydrochloride acts as an intermediate used in organic synthesis. It is also used in the preparation of 3,4-diaminopyridine.
Solubility
Soluble in water.
Notes
Hygroscopic and moisture sensitive.Incompatible with oxidizing agent and strong reducing agents.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 94602-04-7
Molekylær formel C7H9ClN2O3
Molekylvægt (g/mol) 204.61
MDL nummer MFCD01075663
InChI nøgle HNMWMLXQJBWURC-UHFFFAOYSA-N
Synonym 4-ethoxy-3-nitropyridine hydrochloride, 3-nitro-4-ethoxypyridine hydrochloride, 4-ethoxy-3-nitro-pyridine hydrochloride, 4-ethoxy-3-nitropyridine hydrogen chloride, pyridine, 4-ethoxy-3-nitro-, monohydrochloride, 4-ethoxy-3-nitropyridinehydrochloride, pubchem6674, labotest-bb lt00454779, 4-ethoxy-3-nitropyridine hcl, ksc496g1h
PubChem CID 12407497
IUPAC navn 4-ethoxy-3-nitropyridine;hydrochloride
SMIL CCOC1=C(C=NC=C1)[N+](=O)[O-].Cl
Tekniske data
Kemisk navn eller materiale 4-Ethoxy-3-nitropyridine hydrochloride
Smeltepunkt ∼270°C
Mængde 5 g
Beilstein 3712456
Følsomhed Moisture sensitive
Opløselighedsinformation Soluble in water.
Formel vægt 204.61
Procent renhed 97%