missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-fluorobenzeneboronic acid, 98%, Thermo Scientific™

Catalog Number 15439207
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
279263-10-4
C8H10BFO3
183.97
MFCD04115667
ZONJMULNQBNBGU-UHFFFAOYSA-N
4-ethoxy-3-fluorophenyl boronic acid, 4-ethoxy-3-fluorobenzeneboronic acid, 4-ethoxy-3-fluorophenylboronicacid, 3-fluoro-4-ethoxyphenyl boronic acid, boronic acid, 4-ethoxy-3-fluorophenyl, 4-ethoxy-3-fluoro-phenyl boronic acid, acmc-1cp7m, 4-borono-2-fluorophenetole, 3-fluoro-4-ethoxyphenylboronic acid, 4-ethoxy-3-fluorophenyl boronicacid
2782821
(4-ethoxy-3-fluorophenyl)boronic acid
CCOC1=CC=C(C=C1F)B(O)O
This item is not returnable. View return policy
279263-10-4
C8H10BFO3
183.97
MFCD04115667
ZONJMULNQBNBGU-UHFFFAOYSA-N
4-ethoxy-3-fluorophenyl boronic acid, 4-ethoxy-3-fluorobenzeneboronic acid, 4-ethoxy-3-fluorophenylboronicacid, 3-fluoro-4-ethoxyphenyl boronic acid, boronic acid, 4-ethoxy-3-fluorophenyl, 4-ethoxy-3-fluoro-phenyl boronic acid, acmc-1cp7m, 4-borono-2-fluorophenetole, 3-fluoro-4-ethoxyphenylboronic acid, 4-ethoxy-3-fluorophenyl boronicacid
2782821
(4-ethoxy-3-fluorophenyl)boronic acid
CCOC1=CC=C(C=C1F)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 108°C to 113°C
Quantity 1 g
Formula Weight 183.97
Percent Purity 98%
Chemical Name or Material 4-Ethoxy-3-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'