missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3,5-difluorophenylacetic acid, 97%, Thermo Scientific™

Catalog Number 15434537
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1017779-13-3
C10H10F2O3
216.18
MFCD09258681
PVRHYUNNDFVJHY-UHFFFAOYSA-N
3,5-difluoro-4-ethoxyphenylacetic acid, 2-4-ethoxy-3,5-difluorophenyl acetic acid, 4-ethoxy-3,5-difluorophenyl acetic acid, 4-ethoxy-3,5-difluorophenylacetic acid, 4-ethoxy-3,5-difluorophenylacetic acid, jrd
46737600
2-(4-ethoxy-3,5-difluorophenyl)acetic acid
CCOC1=C(F)C=C(CC(O)=O)C=C1F
This item is not returnable. View return policy
1017779-13-3
C10H10F2O3
216.18
MFCD09258681
PVRHYUNNDFVJHY-UHFFFAOYSA-N
3,5-difluoro-4-ethoxyphenylacetic acid, 2-4-ethoxy-3,5-difluorophenyl acetic acid, 4-ethoxy-3,5-difluorophenyl acetic acid, 4-ethoxy-3,5-difluorophenylacetic acid, 4-ethoxy-3,5-difluorophenylacetic acid, jrd
46737600
2-(4-ethoxy-3,5-difluorophenyl)acetic acid
CCOC1=C(F)C=C(CC(O)=O)C=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 55°C to 58°C
Quantity 250 mg
Formula Weight 216.18
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-3,5-difluorophenylacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'