Learn More
4-Ethoxy-2-fluorobenzoyl chloride, 97%, Thermo Scientific™
Product Code. 15426727
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426727 5 g 5g
15416727 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15426727 Supplier Thermo Scientific Alfa Aesar Supplier No. H33851.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1034248-85-5
Molecular Formula C9H8ClFO2
Molecular Weight (g/mol) 202.61
MDL Number MFCD13196594
InChI Key FTJONAPGITZXSQ-UHFFFAOYSA-N
PubChem CID 57361364
IUPAC Name 4-ethoxy-2-fluorobenzoyl chloride
SMILES CCOC1=CC(F)=C(C=C1)C(Cl)=O
Specifications
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 202.61
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2-fluorobenzoyl chloride