missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,6-difluorobenzoic acid, 97%, Thermo Scientific™

Catalog Number p-7032849
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
916483-56-2
C9H8F2O3
202.157
MFCD13196576
CRVNWKFFDWXCGB-UHFFFAOYSA-N
2,6-difluoro-4-ethoxybenzoic acid, benzoic acid, 4-ethoxy-2,6-difluoro
57361345
4-ethoxy-2,6-difluorobenzoic acid
CCOC1=CC(=C(C(=C1)F)C(=O)O)F
This item is not returnable. View return policy
916483-56-2
C9H8F2O3
202.157
MFCD13196576
CRVNWKFFDWXCGB-UHFFFAOYSA-N
2,6-difluoro-4-ethoxybenzoic acid, benzoic acid, 4-ethoxy-2,6-difluoro
57361345
4-ethoxy-2,6-difluorobenzoic acid
CCOC1=CC(=C(C(=C1)F)C(=O)O)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 195°C to 198°C
Quantity 1 g
Formula Weight 202.15
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2,6-difluorobenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'