missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Ethoxy-2,6-difluorobenzaldehyde, 97%
Brand: Alfa Aesar™ H31753.03
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
1017779-48-4 | |
186.158 | |
SHCJRMIWXNHWJF-UHFFFAOYSA-N | |
46737615 | |
CCOC1=CC(=C(C(=C1)F)C=O)F |
C9H8F2O2 | |
MFCD09258695 | |
benzaldehyde, 4-ethoxy-2,6-difluoro, 4-ethoxy-2,6-difluorobenzaldehyde, jrd, 2,6-difluoro-4-ethoxybenzaldehyde | |
4-ethoxy-2,6-difluorobenzaldehyde |
Specifications
4-Ethoxy-2,6-difluorobenzaldehyde | |
70°C to 72°C | |
1017779-48-4 | |
MFCD09258695 | |
SHCJRMIWXNHWJF-UHFFFAOYSA-N | |
4-ethoxy-2,6-difluorobenzaldehyde | |
46737615 | |
97% |
Air Sensitive | |
1g | |
C9H8F2O2 | |
benzaldehyde, 4-ethoxy-2,6-difluoro, 4-ethoxy-2,6-difluorobenzaldehyde, jrd, 2,6-difluoro-4-ethoxybenzaldehyde | |
CCOC1=CC(=C(C(=C1)F)C=O)F | |
186.158 | |
186.16 |