Få mere at vide
4-(Ethenesulfonyl)benzonitrile, TRC
Artikelnummer. 30334604
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30334604 100 mg
30344070 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30334604 Leverandør Toronto Research Chemicals Leverandørnr. TRCB125695100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Ethenesulfonyl)benzonitrile
CAS 1154397-48-4
Molekylær formel C9H7NO2S
Mængde 100 mg
InChI formel InChI=1S/C9H7NO2S/c1-2-13(11,12)9-5-3-8(7-10)4-6-9/h2-6H,1H2
SMIL C=CS(=O)(=O)c1ccc(cc1)C#N
IUPAC navn 4-ethenylsulfonylbenzonitrile
Molekylvægt (g/mol) 193.22
Formel vægt 193.0197