Learn More
4-Epianhydrochlortetracycline hydrochloride, can be used as secondary standard, Thermo Scientific Chemicals
Catalog Number p-125150
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
158018-53-2
C22H21ClN2O7·HCl
497.33
MFCD00143313
ISGAAFMBTIWTEU-PXAZKYFKSA-N
4-epianhydrochlortetracycline hydrochloride, 4-epianhydrochlortetracycline hcl, epianhydrochlortetracycline hydrochloride, 2z,4r,4as,12as-2-amino hydroxy methylidene-7-chloro-4-dimethylamino-10,11,12a-trihydroxy-6-methyl-4a,5-dihydro-4h-tetracene-1,3,12-trione hydrochloride, 4r,4as,12ar-7-chloro-4-dimethylamino-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide-hydrogen chloride 1/1, 4r,4as,12as-7-chloro-4-dimethylamino-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-4a,5-dihydro-4h-tetracene-2-carboxamide hydrochloride, 4r-4, a,4a, a,12a, a-7-chloro-4-dimethylamino-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride
54710413
(4R,4aS,12aR)-7-chloro-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
CC1=C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C14)Cl)O)O)O)O)C(=O)N)N(C)C.Cl
158018-53-2
C22H21ClN2O7·HCl
497.33
MFCD00143313
ISGAAFMBTIWTEU-PXAZKYFKSA-N
4-epianhydrochlortetracycline hydrochloride, 4-epianhydrochlortetracycline hcl, epianhydrochlortetracycline hydrochloride, 2z,4r,4as,12as-2-amino hydroxy methylidene-7-chloro-4-dimethylamino-10,11,12a-trihydroxy-6-methyl-4a,5-dihydro-4h-tetracene-1,3,12-trione hydrochloride, 4r,4as,12ar-7-chloro-4-dimethylamino-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-3,4,4a,5,12,12a-hexahydrotetracene-2-carboxamide-hydrogen chloride 1/1, 4r,4as,12as-7-chloro-4-dimethylamino-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-4a,5-dihydro-4h-tetracene-2-carboxamide hydrochloride, 4r-4, a,4a, a,12a, a-7-chloro-4-dimethylamino-1,4,4a,5,12,12a-hexahydro-3,10,11,12a-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride
54710413
(4R,4aS,12aR)-7-chloro-4-(dimethylamino)-1,10,11,12a-tetrahydroxy-6-methyl-3,12-dioxo-4a,5-dihydro-4H-tetracene-2-carboxamide;hydrochloride
CC1=C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C14)Cl)O)O)O)O)C(=O)N)N(C)C.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 222.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 80% min. (as is,calculated)
Packaging Glass bottle
Quantity 100 mg
Solubility Information Solubility in water: 10g/L. Other solubilities: 29g/L methanol
Formula Weight 497.33
Percent Purity ≥97.0% Total Base (on anhydrous substance), ≥80 wt% (as is, calculated) (HPLC)
Physical Form Powder
Chemical Name or Material 4-Epianhydrochlortetracycline hydrochloride, Can be used as secondary standard
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Epianhydrochlortetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.