Få mere at vide
4-Dodecanolide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | gamma-Dodecalactone |
| CAS | 2305-05-7 |
| Molekylær formel | C12 H22 O2 |
| Mængde | 50 g |
| Synonym | 2(3H)-Furanone, dihydro-5-octyl-, Dodecanoic acid, 4-hydroxy-, γ-lactone (6CI,7CI), Dihydro-5-octyl-2(3H)-furanone, (RS)-γ-Dodecalactone, (±)-4-n-Octylbutyrolactone, (±)-γ-Dodecalactone, 4-Dodecanolide, 4-Hydroxydodecanoic acid lactone, 4-Hydroxydodecanoic acid γ-lactone, 5-Octyltetrahydro-2-furanone, NSC 26511, γ-Dodecalactone, γ-Dodecanolactone, γ-Laurolactone, γ-Octyl-γ-butyrolactone |
| InChI formel | InChI=1S/C12H22O2/c1-2-3-4-5-6-7-8-11-9-10-12(13)14-11/h11H,2-10H2,1H3 |
| SMIL | O=C1OC(CC1)CCCCCCCC |
| IUPAC navn | 5-octyloxolan-2-one |
| Molekylvægt (g/mol) | 198.3 |
| Formel vægt | 198.162 |