missing translation for 'onlineSavingsMsg'
Learn More

4-Diphenylacetoxy-N-methylpiperidine methiodide, Thermo Scientific™

Catalog Number p-7042371
Click to view available options
Quantity:
25 mg
100 mg
This item is not returnable. View return policy
1952-15-4
C21H26INO2
451.348
MFCD00078564
WWJHRSCUAQPFQO-UHFFFAOYSA-M
4-damp methiodide, 4-damp, 4-diphenylacetoxy-n-methylpiperidine methiodide, 4-damp iodide, 1,1-dimethyl-4-diphenylacetoxypiperidinium iodide, 4-diphenylacetyl oxy-1,1-dimethylpiperidinium iodide, piperidinium, 4-diphenylacetyl oxy-1,1-dimethyl-, iodide, 4-diphenylacetyl oxy-1,1-dimethyl-piperidinium iodide, 4-2,2-diphenylacetyl oxy-1,1-dimethylpiperidin-1-ium iodide, chembl76897
3014059
CHEBI:73340
(1,1-dimethylpiperidin-1-ium-4-yl) 2,2-diphenylacetate;iodide
C[N+]1(CCC(CC1)OC(=O)C(C2=CC=CC=C2)C3=CC=CC=C3)C.[I-]
This item is not returnable. View return policy
1952-15-4
C21H26INO2
451.348
MFCD00078564
WWJHRSCUAQPFQO-UHFFFAOYSA-M
4-damp methiodide, 4-damp, 4-diphenylacetoxy-n-methylpiperidine methiodide, 4-damp iodide, 1,1-dimethyl-4-diphenylacetoxypiperidinium iodide, 4-diphenylacetyl oxy-1,1-dimethylpiperidinium iodide, piperidinium, 4-diphenylacetyl oxy-1,1-dimethyl-, iodide, 4-diphenylacetyl oxy-1,1-dimethyl-piperidinium iodide, 4-2,2-diphenylacetyl oxy-1,1-dimethylpiperidin-1-ium iodide, chembl76897
3014059
CHEBI:73340
(1,1-dimethylpiperidin-1-ium-4-yl) 2,2-diphenylacetate;iodide
C[N+]1(CCC(CC1)OC(=O)C(C2=CC=CC=C2)C3=CC=CC=C3)C.[I-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 211°C to 213°C
Color White
Quantity 25 mg
Formula Weight 451.4
Physical Form Powder
Chemical Name or Material 4-Diphenylacetoxy-N-methylpiperidine methiodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'