Learn More
4-(Dimethylamino)pyridine, Sigma-Aldrich™
Product Code. 15866640
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15866640 100 g 100g
15801001 25 g 25g
15811001 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15866640 Supplier Sigma Aldrich Supplier No. 8204990100
 Restricted Item
Forventet forsendelse: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1122-58-3
Grade Research
Molecular Weight (g/mol) 122.17
Specifications
Melting Point 110°C to 112°C
Boiling Point 138°C to 140°C/24 hPa
MDL Number MFCD00006418
Quantity 100 g
Synonym 4-(Dimethylamino)pyridine, 4-(Dimethylamino)pyridine