Få mere at vide
4-(Dimethylamino)benzoic-2,3,5,6-d4 Acid, TRC
Artikelnummer. 30374489
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30374489 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374489 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47091610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Dimethylamino)benzoic-2,3,5,6-d4 Acid
CAS 1175002-07-9
Molekylær formel C9H7D4NO2
Mængde 10 mg
Synonym 4-(N,N-Dimethylamino)benzoic-2,3,5,6-d4 Acid, N,N-Dimethyl-4-aminobenzoic-2,3,5,6-d4 Acid, N,N-Dimethyl-p-aminobenzoic-2,3,5,6-d4 Acid, p-(Dimethylamino)benzoic-2,3,5,6-d4 Acid
InChI formel InChI=1S/C9H11NO2/c1-10(2)8-5-3-7(4-6-8)9(11)12/h3-6H,1-2H3,(H,11,12)/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(C(=O)O)c([2H])c([2H])c1N(C)C
IUPAC navn 2,3,5,6-tetradeuterio-4-(dimethylamino)benzoic acid
Molekylvægt (g/mol) 169.21
Formel vægt 169.1041