missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(Difluoromethoxy)-3-methoxybenzoic acid, 97%

Brand:  Alfa Aesar™ H31594.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15444187

  • 683.00 DKK / 1g

Estimated Shipment
10-03-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

162401-59-4
218.156
QEZBOPWZULAPBJ-UHFFFAOYSA-N
2060451
COC1=C(C=CC(=C1)C(=O)O)OC(F)F
C9H8F2O4
MFCD03964657
4-difluoromethoxy-3-methoxybenzoic acid, 4-difluoromethoxy-3-methoxy-benzoic acid
4-(difluoromethoxy)-3-methoxybenzoic acid

Specifications

4-(Difluoromethoxy)-3-methoxybenzoic acid
1g
C9H8F2O4
4-difluoromethoxy-3-methoxybenzoic acid, 4-difluoromethoxy-3-methoxy-benzoic acid
COC1=C(C=CC(=C1)C(=O)O)OC(F)F
218.156
218.15
169°C to 172°C
162401-59-4
MFCD03964657
QEZBOPWZULAPBJ-UHFFFAOYSA-N
4-(difluoromethoxy)-3-methoxybenzoic acid
2060451
97%
Videos
SDS Documents