missing translation for 'onlineSavingsMsg'
Learn More

4-(Dibutylamino)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-4258934
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
90134-10-4
C15H23NO
233.36
MFCD00075134
VNWPLOWYHIDMEB-UHFFFAOYSA-N
4-dibutylamino benzaldehyde, 4-dibutylamino-benzaldehyde, 4-dibutylaminobenzaldehyd, 4-dibutylaminobenzoic aldehyde
605391
4-(dibutylamino)benzaldehyde
CCCCN(CCCC)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
90134-10-4
C15H23NO
233.36
MFCD00075134
VNWPLOWYHIDMEB-UHFFFAOYSA-N
4-dibutylamino benzaldehyde, 4-dibutylamino-benzaldehyde, 4-dibutylaminobenzaldehyd, 4-dibutylaminobenzoic aldehyde
605391
4-(dibutylamino)benzaldehyde
CCCCN(CCCC)C1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Boiling Point 148.0°C (1.0 Torr)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 233.35
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(Dibutylamino)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Dibutylamino)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'