Få mere at vide
4-Despiperidinyl-4-hydroxy Dipyridamole, TRC
Artikelnummer. 30367100
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367100 10 mg
30354124 5 mg
30360864 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30367100 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29720010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 68006-07-5
Molekylær formel C19H31N7O5
Molekylvægt (g/mol) 437.49
Synonym 2,6-Bis[bis(2-hydroxyethyl)amino]-8-(1-piperidinyl)pyrimido[5,4-d]pyrimidin-4(3H)-one
SMIL OCCN(CCO)C1=Nc2c(nc(nc2C(=O)N1)N(CCO)CCO)N3CCCCC3
Tekniske data
Kemisk navn eller materiale 4-Despiperidinyl-4-hydroxy Dipyridamole
Mængde 10 mg
InChI formel InChI=1S/C19H31N7O5/c27-10-6-25(7-11-28)18-21-15-14(16(22-18)24-4-2-1-3-5-24)20-19(23-17(15)31)26(8-12-29)9-13-30/h27-30H,1-13H2,(H,20,23,31)
Formel vægt 437.2387