Få mere at vide
4-Desmethyl Flucarbazone Sodium Salt, TRC
Artikelnummer. 30361048
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361048 100 mg
30353081 50 mg
30350909 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30361048 Leverandør Toronto Research Chemicals Leverandørnr. TRCD343230100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 328385-26-8
Molekylær formel C11H9F3N4NaO6S
Molekylvægt (g/mol) 404.25
Synonym 2,5-Dihydro-3-methoxy-5-oxo-N-[[2-(trifluoromethoxy)phenyl]sulfonyl]-1H-1,2,4-triazole-1-carboxamide Sodium Salt
IUPAC navn sodium;(3-methoxy-5-oxo-4H-1,2,4-triazole-1-carbonyl)-[2-(trifluoromethoxy)phenyl]sulfonylazanide
SMIL [Na+].COC1=NN(C(=O)[N-]S(=O)(=O)c2ccccc2OC(F)(F)F)C(=O)N1
Tekniske data
Kemisk navn eller materiale 4-Desmethyl Flucarbazone Sodium Salt
Mængde 100 mg
InChI formel InChI=1S/C11H9F3N4O6S.Na/c1-23-8-15-9(19)18(16-8)10(20)17-25(21,22)7-5-3-2-4-6(7)24-11(12,13)14;/h2-5H,1H3,(H2,15,16,17,19,20);/q;+1/p-1
Formel vægt 404.0014