Få mere at vide
4-Desmethoxy Omeprazole-d3, TRC
Artikelnummer. 30368047
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30368047 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368047 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29056210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Desmethoxy Omeprazole-d3
CAS 1794759-05-9
Molekylær formel C16H14D3N3O2S
Mængde 10 mg
Synonym 2-[[(3,5-Dimethyl-2-pyridinyl)methyl]sulfinyl]-6-(methoxy-d3)-1H-benzimidazole, H 180/29-d3
InChI formel InChI=1S/C16H17N3O2S/c1-10-6-11(2)15(17-8-10)9-22(20)16-18-13-5-4-12(21-3)7-14(13)19-16/h4-8H,9H2,1-3H3,(H,18,19)/i3D3
SMIL [2H]C([2H])([2H])Oc1ccc2nc([nH]c2c1)S(=O)Cc3ncc(C)cc3C
IUPAC navn 2-[(3,5-dimethylpyridin-2-yl)methylsulfinyl]-6-(trideuteriomethoxy)-1H-benzimidazole
Molekylvægt (g/mol) 318.41
Formel vægt 318.123