Få mere at vide
4-Desmethoxy Omeprazole-d3 Sulfide, TRC
Artikelnummer. 30358556
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30358556 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358556 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29056725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Desmethoxy Omeprazole-d3 Sulfide
CAS 1794938-78-5
Molekylær formel C16H14D3N3OS
Mængde 25 mg
Synonym 2-[[(3,5-Dimethyl-2-pyridinyl)methyl]thio]-6-methoxy-1H-benzimidazole-d3
InChI formel InChI=1S/C16H17N3OS/c1-10-6-11(2)15(17-8-10)9-21-16-18-13-5-4-12(20-3)7-14(13)19-16/h4-8H,9H2,1-3H3,(H,18,19)/i3D3
SMIL [2H]C([2H])([2H])Oc1ccc2nc(SCc3ncc(C)cc3C)[nH]c2c1
IUPAC navn 2-[(3,5-dimethylpyridin-2-yl)methylsulfanyl]-6-(trideuteriomethoxy)-1H-benzimidazole
Molekylvægt (g/mol) 302.41
Formel vægt 302.1281