Få mere at vide
4-Desethoxy-2-ethoxy Dapagliflozin, >90%, TRC
Artikelnummer. 30348847
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30348847 5 mg
30359016 25 mg
30364498 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30348847 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4332685MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2040305-05-1
Molekylær formel C21H25ClO6
Molekylvægt (g/mol) 408.87
Synonym 2S,3R,4R,5S,6R)-2-(4-Chloro-3-(2-ethoxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, Dapagliflozin Ortho Isomer
IUPAC navn (2S,3R,4R,5S,6R)-2-[4-chloro-3-[(2-ethoxyphenyl)methyl]phenyl]-6-(hydroxymethyl)oxane-3,4,5-triol
SMIL CCOc1ccccc1Cc2cc(ccc2Cl)[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O
Tekniske data
Kemisk navn eller materiale 2S,3R,4R,5S,6R-2-(4-Chloro-3-(2-ethoxybenzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
Mængde 5 mg
InChI formel InChI=1S/C21H25ClO6/c1-2-27-16-6-4-3-5-12(16)9-14-10-13(7-8-15(14)22)21-20(26)19(25)18(24)17(11-23)28-21/h3-8,10,17-21,23-26H,2,9,11H2,1H3/t17-,18-,19+,20-,21+/m1/s1
Formel vægt 408.134