Få mere at vide
4-Cyclohexylbutanoic Acid, Mikromol™
Beskrivelse
4-Cyclohexylbutanoic Acid
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 4-cyclohexylbutanoic acid |
| CAS | 4441-63-8 |
| Analyt- eller komponentnavne | 4-cyclohexylbutanoic acid |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C10 H18 O2 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Sodium Phenylbutyrate Imp. C (EP), Phenylbutyrate USP Related Compound C, Phenylbutyrate USP RC C, Cyclohexanebutanoic acid, Cyclohexanebutyric acid, 4-Cyclohexylbutanoic acid, 4-Cyclohexylbutyric acid, NSC 2158, NSC 406925, Phenylbutyrate Related Compound C, Sodium Phenylbutyrate Impurity C |
| InChI formel | InChI=1S/C10H18O2/c11-10(12)8-4-7-9-5-2-1-3-6-9/h9H,1-8H2,(H,11,12) |
| SMIL | OC(=O)CCCC1CCCCC1 |
| IUPAC navn | 4-cyclohexylbutanoic acid |
| Vis mere |