missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanophenol, 99%, Thermo Scientific Chemicals

Catalog Number p-3778009
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
767-00-0
C7H5NO
119.12
MFCD00002312
CVNOWLNNPYYEOH-UHFFFAOYSA-N
4-cyanophenol, p-hydroxybenzonitrile, p-cyanophenol, benzonitrile, 4-hydroxy, benzonitrile, p-hydroxy, 4-cyano phenol, 4-hydroxybenzoic acid nitrile, 4-hydroxy-benzonitrile, 4-hydroxy benzonitrile, 4-hydroxybenzenecarbonitrile
13019
CHEBI:38622
4-hydroxybenzonitrile
C1=CC(=CC=C1C#N)O
This item is not returnable. View return policy
767-00-0
C7H5NO
119.12
MFCD00002312
CVNOWLNNPYYEOH-UHFFFAOYSA-N
4-cyanophenol, p-hydroxybenzonitrile, p-cyanophenol, benzonitrile, 4-hydroxy, benzonitrile, p-hydroxy, 4-cyano phenol, 4-hydroxybenzoic acid nitrile, 4-hydroxy-benzonitrile, 4-hydroxy benzonitrile, 4-hydroxybenzenecarbonitrile
13019
CHEBI:38622
4-hydroxybenzonitrile
C1=CC(=CC=C1C#N)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 110°C to 113°C
Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NCC6H4OH
Quantity 25 g
Beilstein 10, 167
Solubility Information Solubility in water: slightly soluble
Formula Weight 119.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyanophenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyanophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'