Få mere at vide
4-Cyano-4'-n-pentyl-d11-biphenyl, CDN
Artikelnummer. 30318220
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318220 0.01 g
30313503 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318220 Leverandør CDN Leverandørnr. CDND20980.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Cyano-4'-pentylbiphenyl-D11 (pentyl-D11)
CAS 57078-20-3
Molekylær formel C18 D11 H8 N
Mængde 0.01 g
Synonym [1,1'-Biphenyl]-4-carbonitrile, 4'-(pentyl-1,1,2,2,3,3,4,4,5,5,5-d11)- (ACI), 4'-(Pentyl-1,1,2,2,3,3,4,4,5,5,5-d11)[1,1'-biphenyl]-4-carbonitrile (ACI), [1,1'-Biphenyl]-4-carbonitrile, 4'-(pentyl-d11)- (9 CI), 4-Cyano-4'-pentyl-d11-biphenyl, 4-Pentyl-d11-4'-cyanobiphenyl, 4-Cyano-4'-pentylbiphenyl-D11 (pentyl-D11), 4-Cyano-4'-pentylbiphenyl-D11 (pentyl-D11), 4-Cyano-4'-pentylbiphenyl-D11 (pentyl-D11)
InChI formel InChI=1 S/C18H19N/c1-2-3-4-5-15-6-10-17(11-7-15)18-12-8-16(14-19)9-13-18/h6-13 H,2-5H2,1H3/i1D3,2D2,3D2,4D2,5D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])c1ccc(cc1)c2ccc(cc2)C#N
IUPAC navn 4-[4-(1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentyl)phenyl]benzonitrile
Molekylvægt (g/mol) 260.4179
Formel vægt 260.2208 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity