Få mere at vide
4-Cyano-4-[(ethylsulfanylthiocarbonyl)sulfanyl]pentanoic Acid, TRC
Artikelnummer. 30357624
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30357624 100 mg
30355683 10 mg
30357833 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30357624 Leverandør Toronto Research Chemicals Leverandørnr. TRCC998578100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1137725-46-2
Molekylær formel C9H13NO2S3
Molekylvægt (g/mol) 263.4
IUPAC navn 4-cyano-4-ethylsulfanylcarbothioylsulfanylpentanoic acid
SMIL CCSC(=S)SC(C)(CCC(=O)O)C#N
Tekniske data
Kemisk navn eller materiale 4-Cyano-4-[(ethylsulfanylthiocarbonyl)sulfanyl]pentanoic Acid
Mængde 100 mg
InChI formel InChI=1S/C9H13NO2S3/c1-3-14-8(13)15-9(2,6-10)5-4-7(11)12/h3-5H2,1-2H3,(H,11,12)
Formel vægt 263.0108