missing translation for 'onlineSavingsMsg'
Learn More
4'-Cyano-4-biphenylyl trans-4-ethylcyclohexanecarboxylate, 97%, Thermo Scientific™ Product Code.: 15421906

4'-Cyano-4-biphenylyl trans-4-ethylcyclohexanecarboxylate, 97%, Thermo Scientific™

Product Code. 15421906
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15421906

Brand: Thermo Scientific Alfa Aesar H55054.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67284-56-4
Molecular Formula C22H23NO2
Molecular Weight (g/mol) 333.431
MDL Number MFCD09751067
InChI Key OYZITXKNUGHHKV-UHFFFAOYSA-N
Synonym trans-4'-cyano-1,1'-biphenyl-4-yl 4-ethylcyclohexanecarboxylate, trans-4'-cyano 1,1'-biphenyl-4-yl 4-ethylcyclohexanecarboxylate, 4-4-cyanophenyl phenyl 4-ethylcyclohexane-1-carboxylate, 4'-cyano-1,1'-biphenyl-4-yl 1s,4r-4-ethylcyclohexane-1-carboxylate, 4-cyano-4'-biphenyl 4-ethyl-trans-cyclohexanecarboxylate, 4'-cyanobiphenyl-4-yl trans-4-ethyl-cyclohexanecarboxylate, cyclohexanecarboxylic acid, 4-ethyl-, 4'-cyano 1,1'-biphenyl-4-yl ester, trans, 4-cyanobiphenyl-4'-trans-ethylcyclohexylcarboxylate, 4-4-cyanophenyl phenyl 4-ethylcyclohexanecarboxylate
PubChem CID 171826
IUPAC Name [4-(4-cyanophenyl)phenyl] 4-ethylcyclohexane-1-carboxylate
SMILES CCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)C3=CC=C(C=C3)C#N

Specifications

Melting Point 83°C to 86°C
Quantity 1 g
Formula Weight 333.43
Percent Purity 97%
Chemical Name or Material 4'-Cyano-4-biphenylyl trans-4-ethylcyclohexanecarboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Product Code
Lot NumberBE010501 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4'-Cyano-4-biphenylyl trans-4-ethylcyclohexanecarboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.