missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-methoxybenzeneboronic acid, 96%, Thermo Scientific™

Catalog Number 15477788
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1256345-67-1
C8H8BNO3
176.966
CCUBWDNQHCIXNF-UHFFFAOYSA-N
4-cyano-2-methoxyphenyl boronic acid, 4-cyano-2-methoxyphenyl boronicacid, boronic acid, b-4-cyano-2-methoxyphenyl, acmc-209avk, 4-cyano-2-methoxy-phenyl boronic acid
53216296
(4-cyano-2-methoxyphenyl)boronic acid
B(C1=C(C=C(C=C1)C#N)OC)(O)O
This item is not returnable. View return policy
1256345-67-1
C8H8BNO3
176.966
CCUBWDNQHCIXNF-UHFFFAOYSA-N
4-cyano-2-methoxyphenyl boronic acid, 4-cyano-2-methoxyphenyl boronicacid, boronic acid, b-4-cyano-2-methoxyphenyl, acmc-209avk, 4-cyano-2-methoxy-phenyl boronic acid
53216296
(4-cyano-2-methoxyphenyl)boronic acid
B(C1=C(C=C(C=C1)C#N)OC)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 176.97
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Cyano-2-methoxybenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'