missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number p-7039698
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1035235-29-0
C13H15BFNO2
247.08
MFCD09998162
FEUINZSEHIJUBU-UHFFFAOYSA-N
3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-2-fluorophenylboronic acid, pinacol ester, 4-cyano-2-fluorophenylboronic acid pinacol ester, 4-cyano-2-fluorobenzeneboronic acid pinacol ester, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-2-fluorophenylboronicacidpinacolester, 4-cyano-2-fluorobenzeneboronic acid, pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 3-fluoro-4-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl benzonitrile, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl benzenecarbonitrile
44755210
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C#N
This item is not returnable. View return policy
1035235-29-0
C13H15BFNO2
247.08
MFCD09998162
FEUINZSEHIJUBU-UHFFFAOYSA-N
3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-2-fluorophenylboronic acid, pinacol ester, 4-cyano-2-fluorophenylboronic acid pinacol ester, 4-cyano-2-fluorobenzeneboronic acid pinacol ester, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-2-fluorophenylboronicacidpinacolester, 4-cyano-2-fluorobenzeneboronic acid, pinacol ester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 3-fluoro-4-4,4,5,5-tetramethyl-1,3-dioxaborolan-2-yl benzonitrile, 3-fluoro-4-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl benzenecarbonitrile
44755210
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 68°C to 70°C
Quantity 250 mg
Formula Weight 247.08
Percent Purity 96%
Chemical Name or Material 4-Cyano-2-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'