Få mere at vide
4-Chlorothiobenzamide-2,3,5,6-d4, CDN
Artikelnummer. 30317218
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317218 0.05 g -
30316491 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317218 Leverandør CDN Leverandørnr. CDND65730.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chlorothiobenzamide-2,3,5,6-D4
CAS 1219802-58-0
Molekylær formel C7 D4 H2 Cl N S
Mængde 0.05 g
Synonym 4-Chlorothiobenzamide-2,3,5,6-D4, 4-Chlorobenzene-2,3,5,6-d4-carbothioamide, 4-Chlorobenzothiamide-d4
InChI formel InChI=1 S/C7H6ClNS/c8-6-3-1-5(2-4-6)7(9)10/h1-4 H,(H2,9,10)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(C(=S)N)c([2 H])c([2 H])c1Cl
IUPAC navn 4-chloro-2,3,5,6-tetradeuteriobenzenecarbothioamide
Molekylvægt (g/mol) 175.6719
Formel vægt 175.0161 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity