Learn More
4-(Chlorosulfonyl)benzoic acid, 96%, Thermo Scientific Chemicals
Catalog Number p-4509520
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
10130-89-9
C7H5ClO4S
220.63
PTCSSXYPZOFISK-UHFFFAOYSA-N
4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811
82385
4-chlorosulfonylbenzoic acid
C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl
10130-89-9
C7H5ClO4S
220.63
PTCSSXYPZOFISK-UHFFFAOYSA-N
4-chlorosulfonyl benzoic acid, benzoic acid, 4-chlorosulfonyl, p-chlorosulfonyl benzoic acid, 4-chlorosulphonyl benzoic acid, p-carboxybenzenesulfonyl chloride, benzoic acid, p-chlorosulfonyl, 4-chlorosulfonyl-benzoic acid, 4-carboxybenzenesulfonyl chloride, 4-carboxy-benzenesulfonylchloride, pubchem14811
82385
4-chlorosulfonylbenzoic acid
C1=CC(=CC=C1C(=O)O)S(=O)(=O)Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 230.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CO2HC6H4SO2Cl
Quantity 5 g
Solubility Information Solubility in water: decomposes.
Formula Weight 220.63
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-(Chlorosulfonyl)benzoic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-(Chlorosulfonyl)benzoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.