Learn More
4-chlorosalicylic acid, 98%, ACROS Organics™
Brand: Acros Organics 179840250
Additional Details : CAS Number : 5106-98-9 Weight : 0.07500kg
Packaging | Plastic bottle |
---|---|
Quantity | 25g |
Chemical Identifiers
5106-98-9 | |
172.57 | |
LWXFCZXRFBUOOR-UHFFFAOYSA-N | |
78782 | |
C1=CC(=C(C=C1Cl)O)C(=O)O |
C7H5ClO3 | |
MFCD00002449 | |
4-chlorosalicylic acid, 4-chloro salicylic acid, benzoic acid, 4-chloro-2-hydroxy, 2-hydroxy-4-chlorobenzoic acid, 4-chloro-2-hydroxy benzoic acid, 4-chloro-2-hydroxy-benzoic acid, salicylic acid, 4-chloro, 4-chlorosalicylicacid, pubchem3845, 4-chloro-salicylic acid | |
4-chloro-2-hydroxybenzoic acid |
Specifications
4-chlorosalicylic acid | |
Plastic bottle | |
209°C to 213°C | |
5106-98-9 | |
100.0 | |
C7H5ClO3 | |
MFCD00002449 | |
4-chlorosalicylic acid, 4-chloro salicylic acid, benzoic acid, 4-chloro-2-hydroxy, 2-hydroxy-4-chlorobenzoic acid, 4-chloro-2-hydroxy benzoic acid, 4-chloro-2-hydroxy-benzoic acid, salicylic acid, 4-chloro, 4-chlorosalicylicacid, pubchem3845, 4-chloro-salicylic acid | |
LWXFCZXRFBUOOR-UHFFFAOYSA-N | |
4-chloro-2-hydroxybenzoic acid | |
78782 | |
Powder |
Authentic | |
White to Beige | |
25g | |
97.5 | |
98% | |
ClC6H3(OH)CO2H | |
10, 101 | |
Solubility in water: slightly soluble in water. Other solubilities: soluble in most organic solvents | |
C1=CC(=C(C=C1Cl)O)C(=O)O | |
172.57 | |
172.57 | |
98% |
Safety and Handling
- 4-Chlorosalicylic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.