missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.
Solubility
Soluble in hot water, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4-Chlorophenylhydrazine hydrochloride is used as a pharmaceutical intermediates.
Solubility
Soluble in hot water, methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Melting Point | ∼216°C (decomposition) |
Odor | Odorless |
Quantity | 5 g |
Beilstein | 3596254 |
Solubility Information | Soluble in hot water,methanol. |
Formula Weight | 179.05 |
Percent Purity | 97% |
Chemical Name or Material | 4-Chlorophenylhydrazine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'