Få mere at vide
4-Chlorophenyl Phenyl-d5 Ether, CDN
Artikelnummer. 30316918
Skift visning
Mængde:
0.1 g
Pakningsstørrelse:
0.1g
Artikelnummer. Mængde unitSize
30316918 0.1 g 0.1g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316918 Leverandør CDN Leverandørnr. CDND23120.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Chlorodiphenyl ether 2',3',4',5',6'-d5
CAS 93951-85-0
Molekylær formel C12 2H5 H4 Cl O
Mængde 0.1 g
Synonym Benzene-d5, (4-chlorophenoxy)- (9 CI), 1-Chloro-4-üentadeuteriophenoxybenzene, 4-Chlorodiphenyl ether (2',3',4',5',6'-D5), 4-Chlorophenyl perdeuteriophenyl ether, CDE-(2',3',4',5',6'-D5) 3;PCDE 3 (2',3',4',5',6'-D5), p-Chlorophenyl perdeuteriophenyl ether, p-Pentadeuteriophenoxychlorobenzene, 4-Chlorophenyl phenyl ether phenyl-d5
InChI formel InChI=1 S/C12H9ClO/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9 H/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(Oc2ccc(Cl)cc2)c([2 H])c1[2 H]
IUPAC navn 1-(4-chlorophenoxy)-2,3,4,5,6-pentadeuteriobenzene
Molekylvægt (g/mol) 209.68
Formel vægt 209.0656 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity