Få mere at vide
(4-Chlorophenyl)guanidine Hydrochloride, Mikromol™
Beskrivelse
(4-Chlorophenyl)guanidine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | (4-Chlorophenyl)guanidine Hydrochloride |
| CAS | 14279-91-5 |
| Analyt- eller komponentnavne | (4-Chlorophenyl)guanidine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C7 H8 Cl N3 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | (4-Chlorophenyl)guanidine Hydrochloride, Guanidine, N-(4-chlorophenyl)-, hydrochloride (1:1), Guanidine, (4-chlorophenyl)-, monohydrochloride (9CI), Guanidine, (p-chlorophenyl)-, monohydrochloride (8CI), 4-Chlorophenylguanidine hydrochloride, BW 65-100, p-Chlorophenyl urea hydrochloride, Chlorhexidine Diacetate Impurity E as Hydrochloride, Chlorhexidine Impurity E as Hydrochloride |
| InChI formel | InChI=1S/C7H8ClN3.ClH/c8-5-1-3-6(4-2-5)11-7(9)10;/h1-4H,(H4,9,10,11);1H |
| SMIL | Cl.NC(=N)Nc1ccc(Cl)cc1 |
| IUPAC navn | 1-(4-chlorophenyl)guanidine;hydrochloride |
| Vis mere |