Få mere at vide
(+/-)-4-Chlorophenyl-5-[(3,4-isopropylidine)-2-methylpyridine]methanol-d4, TRC
Artikelnummer. 30356372
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30356372 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30356372 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3785522.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (+/-)-4-Chlorophenyl-5-[(3,4-isopropylidine)-2-methylpyridine]methanol-d4
CAS 1216968-24-9
Molekylær formel C17H14D4ClNO3
Mængde 2.5 mg
Synonym (+/-)-alpha-(4-Chlorophenyl)-2,2,8-trimethyl-4H-1,3-dioxino[4,5-c]pyridine-5-methanol-d4
InChI formel InChI=1S/C17H18ClNO3/c1-10-16-14(9-21-17(2,3)22-16)13(8-19-10)15(20)11-4-6-12(18)7-5-11/h4-8,15,20H,9H2,1-3H3/i4D,5D,6D,7D
SMIL [2H]c1c([2H])c(C(O)c2cnc(C)c3OC(C)(C)OCc23)c([2H])c([2H])c1Cl
IUPAC navn (4-chloro-2,3,5,6-tetradeuteriophenyl)-(2,2,8-trimethyl-4H-[1,3]dioxino[4,5-c]pyridin-5-yl)methanol
Molekylvægt (g/mol) 323.81
Formel vægt 323.1226