missing translation for 'onlineSavingsMsg'
Få mere at vide

(4-Chlorophenyl)(1-hydroxycyclopentyl)methanone, TRC

Product Code. 30355109
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30355109 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30355109 Supplier Toronto Research Chemicals Supplier No. TRCC38074025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale (4-Chlorophenyl)(1-hydroxycyclopentyl)methanone
CAS 1696839-44-7
Molekylær formel C12H13ClO2
Mængde 25 mg
InChI formel InChI=1S/C12H13ClO2/c13-10-5-3-9(4-6-10)11(14)12(15)7-1-2-8-12/h3-6,15H,1-2,7-8H2
SMIL OC1(CCCC1)C(=O)c2ccc(Cl)cc2
IUPAC navn (4-chlorophenyl)-(1-hydroxycyclopentyl)methanone
Molekylvægt (g/mol) 224.68
Formel vægt 224.0604
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.